missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-proline, 98+%

Product Code. 11339729
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339729 5 g 5g
11319729 50 g 50g
11329729 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339729 Supplier Thermo Scientific Alfa Aesar Supplier No. B21672.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1148-11-4 | C13H15NO4 | 249.266 g/mol

N-(Benzyloxycarbonyl)-L-proline is a potent inhibitor of prolidase; a specific peptidase that cleaves dipeptides with a C-terminal prolyl and hydroxylprolyl residue. It is also used in the synthesis of N-(L-Prolyl)-β-alanine which is a derivative of the naturally occurring beta amino acid β-Alanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Benzyloxycarbonyl)-L-proline is a potent inhibitor of prolidase; a specific peptidase that cleaves dipeptides with a C-terminal prolyl and hydroxylprolyl residue. It is also used in the synthesis of N-(L-Prolyl)-β-alanine which is a derivative of the naturally occurring beta amino acid β-Alanine.

Solubility
Solubility in methanol, almost transparency.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1148-11-4
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD00003170
InChI Key JXGVXCZADZNAMJ-NSHDSACASA-N
Synonym z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline
PubChem CID 101987
IUPAC Name (2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 75°C to 77°C
Quantity 5 g
Beilstein 88579
Optical Rotation −60° (c=1 in Acetic Acid)
Formula Weight 249.27
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-L-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.