missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-glutamic acid 5-benzyl ester, 95%, Thermo Scientific™

Product Code. p-7041460
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15497126

Brand: Thermo Scientific Alfa Aesar H62181.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Benzyloxycarbonyl-L-glutamic acid 5-benzyl ester is used as pharmaceutical intermediates.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5680-86-4
Molecular Formula C20H21NO6
Molecular Weight (g/mol) 371.389
MDL Number MFCD00065697
InChI Key TWIVXHQQTRSWGO-KRWDZBQOSA-N
Synonym z-glu obzl-oh, cbz-glu obzl-oh, 2s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, 5-benzyl n-carbobenzoxy-l-glutamate, n-cbz-l-glutamic acid 5-benzyl ester, s-2-benzyloxycarbonylamino-pentanedioic acid 5-benzyl ester, s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, z-glu obzl oh, pubchem14964, z-glu-obzl-oh
PubChem CID 3049860
IUPAC Name (2S)-5-oxo-5-phenylmethoxy-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Quantity 5 g
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 371.38
Percent Purity 95%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 5-benzyl ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.