missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, 95%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.
Solubility
Soluble in methanol, dimethyl sulfoxide. Slightly soluble in water.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.
Solubility
Soluble in methanol, dimethyl sulfoxide. Slightly soluble in water.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 98°C to 102°C |
| Quantity | 25 g |
| Solubility Information | Soluble in methanol,dimethyl sulfoxide. Slightly soluble in water. |
| Formula Weight | 371.38 |
| Percent Purity | 95% |
| Chemical Name or Material | N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction