missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals

Product Code. p-7040157
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15416268

Brand: Thermo Scientific Alfa Aesar H59838.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Soluble in DMSO, and methanol.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5545-52-8
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00042857
InChI Key HLSLRFBLVZUVIE-LBPRGKRZSA-N
Synonym z-asp otbu-oh, n-cbz-l-aspartic acid 4-tert-butyl ester, cbz-l-aspartic acid 4-tert-butyl ester, cbz-asp obut-oh, z-asp otbu-oh.dcha, cbz-l-aspartic 4-tert-butyl ester, s-2-benzyloxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, z-l-aspartic acid 4-tert-butyl ester, 4-tert-butyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, z-asp otbu-oh.h2o
PubChem CID 111082
IUPAC Name (2S)-4-[(2-methylpropan-2-yl)oxy]-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Linear Formula (CH3)3CO2CCH2CH(NHCO2CH2C6H5)CO2H
Quantity 1 g
Beilstein 2569846
Solubility Information Soluble in DMSO,and methanol.
Formula Weight 323.34
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.