missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-aspartic acid 4-methyl ester, 98%, Thermo Scientific Chemicals

Product Code. 15428426
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3160-47-2
Molecular Formula C13H15NO6
Molecular Weight (g/mol) 281.264
MDL Number MFCD00133585
InChI Key PHMBNDDHIBIDRQ-JTQLQIEISA-N
Synonym z-asp ome-oh, s-2-n-cbz-amino-succinic acid 4-methyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-methyl ester, s-2-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, cbz-l-aspartic acid 4-methyl ester, n-cbz-l-aspartate, pubchem14062, z-l-aspartic acid 4-methyl ester, z-asp ome-oh tlc
PubChem CID 7014888
IUPAC Name (2S)-4-methoxy-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES COC(=O)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Color White
Beilstein 6279422
Formula Weight 281.26
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'