missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-alaninol

Product Code. 15473597
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473597 5 g 5g
15463597 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473597 Supplier Thermo Scientific Alfa Aesar Supplier No. H27066.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66674-16-6 | C11H15NO3 | 209.25 g/mol

N-Benzyloxycarbonyl-L-alaninol is used as an important organic intermediate. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-alaninol is used as an important organic intermediate. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66674-16-6
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00672530
InChI Key AFPHMHSLDRPUSM-VIFPVBQESA-N
Synonym cbz-l-alaninol, s-benzyl 1-hydroxypropan-2-yl carbamate, n-z-l-alaninol, s-benzyl 1-hydroxypropan-2-ylcarbamate, benzyl n-2s-1-hydroxypropan-2-yl carbamate, carbamic acid, 1s-2-hydroxy-1-methylethyl-, phenylmethyl ester, phenylmethyl n-2s-1-oxidanylpropan-2-yl carbamate, n-2s-1-hydroxypropan-2-yl carbamic acid phenylmethyl ester, n-cbz-l-alaninol, cbz-l-ala-ol
PubChem CID 10262499
IUPAC Name benzyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
SMILES C[C@@H](CO)NC(=O)OCC1=CC=CC=C1

Specifications

Solubility Information Soluble in ethanol.
Quantity 5 g
Formula Weight 209.25
Chemical Name or Material N-Benzyloxycarbonyl-L-alaninol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.