missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-alanine, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | N-Benzyloxycarbonyl-L-alanine |
| CAS | 1142-20-7 |
| Molecular Formula | C11 H13 N O4 |
| Quantity | 25 g |
| Synonym | L-Alanine, N-[(phenylmethoxy)carbonyl]-, Alanine, N-carboxy-, N-benzyl ester, L- (8CI), L-Alanine, N-carboxy-, N-benzyl ester (6CI,7CI), N-[(Phenylmethoxy)carbonyl]-L-alanine, (2S)-2-[[(Benzyloxy)carbonyl]amino]propanoic acid, (S)-2-(Benzyloxycarbonylamino)propanoic acid, (S)-2-[(Benzyloxycarbonyl)amino]propionic acid, Benzyloxycarbonyl-L-alanine, Cbz-L-Ala-OH, L-Carbobenzoxyalanine, L-Carbobenzyloxyalanine, L-Cbz alanine, L-N-(Benzyloxycarbonyl)alanine, N-(Benzyloxycarbonyl)-(S)-alanine, N-(Carbobenzoxy)-L-alanine, N-Benzoxycarbonyl-L-alanine, N-Carbobenzyloxy-L-alanine, N-[(Benzyloxy)carbonyl]-L-alanine, NSC 156978, NSC 88469, CBZ-L-alanine |
| InChI Formula | InChI=1S/C11H13NO4/c1-8(10(13)14)12-11(15)16-7-9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,12,15)(H,13,14)/t8-/m0/s1 |
| SMILES | C[C@H](NC(=O)OCc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-2-(phenylmethoxycarbonylamino)propanoic acid |
| Molecular Weight (g/mol) | 223.23 |
| Formula Weight | 223.0845 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction