missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyl)hydroxylamine, 98+%

Product Code. 11357956
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357956 25 g 25g
11347956 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357956 Supplier Thermo Scientific Alfa Aesar Supplier No. L13175.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3426-71-9 | C8H9NO3 | 167.16 g/mol

N-(Benzyloxycarbonyl)hydroxylamine reacts with cyclohepta-1,3-diene to produce 6-oxa-7-aza-bicyclo[3.2.2]non-8-ene-7-carboxylic acid benzyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Benzyloxycarbonyl)hydroxylamine reacts with cyclohepta-1,3-diene to produce 6-oxa-7-aza-bicyclo[3.2.2]non-8-ene-7-carboxylic acid benzyl ester.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with strong oxidizing agents. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3426-71-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00010642
InChI Key PQBSPTAPCMSZAA-UHFFFAOYSA-N
Synonym benzyl hydroxycarbamate, n-benzyloxycarbonyl hydroxylamine, n-carbobenzoxyhydroxylamine, carbamic acid, hydroxy-, phenylmethyl ester, n-benzyloxycarbonylhydroxylamine, benzylhydroxycarbamate, ccris 5076, carbamic acid, hydroxy-, benzyl ester, 1-06-00-00221 beilstein handbook reference, carbamic acid, n-hydroxy-, phenylmethyl ester
PubChem CID 18907
IUPAC Name benzyl N-hydroxycarbamate
SMILES ONC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 65°C to 70°C
Quantity 25 g
Beilstein 1074034
Sensitivity Moisture sensitive
Formula Weight 167.17
Percent Purity ≥98%
Chemical Name or Material N-(Benzyloxycarbonyl)hydroxylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.