missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Benzyloxycarbonyl)ethanolamine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | N-(Benzyloxycarbonyl)ethanolamine |
| CAS | 77987-49-6 |
| Molecular Formula | C10 H13 N O3 |
| Quantity | 10 g |
| Synonym | (2-Hydroxyethyl)carbamic acid benzyl ester, (2-Hydroxyethyl)carbamic acid phenylmethyl ester, 2-(Benzyloxycarbonylamino)ethanol, 2-[N-(Benzyloxycarbonyl)amino]ethanol, Benzyl (2-hydroxyethyl)carbamate, Benzyl N-(2-hydroxyethyl)carbamate, N-(2-Hydroxyethyl)carbamic acid benzyl ester, N-(Benzyloxycarbonyl)ethanolamine, N-Benzyloxycarbonyl-2-aminoethanol, NSC 132101 |
| InChI Formula | InChI=1S/C10H13NO3/c12-7-6-11-10(13)14-8-9-4-2-1-3-5-9/h1-5,12H,6-8H2,(H,11,13) |
| SMILES | OCCNC(=O)OCc1ccccc1 |
| IUPAC Name | benzyl N-(2-hydroxyethyl)carbamate |
| Molecular Weight (g/mol) | 195.215 |
| Formula Weight | 195.09 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction