missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-D-valine, 98+%, Thermo Scientific Chemicals
£21.80 - £60.10
Chemical Identifiers
CAS | 1685-33-2 |
---|---|
Molecular Formula | C13H17NO4 |
Molecular Weight (g/mol) | 251.28 |
MDL Number | MFCD00065703 |
InChI Key | CANZBRDGRHNSGZ-LLVKDONJSA-N |
Synonym | z-d-val-oh, cbz-d-valine, n-benzyloxycarbonyl-d-valine, cbz-d-val-oh, n-cbz-d-valine, z-d-valine, n-carbobenzoxy-d-valine, benzyloxycarbonyl-d-valine, r-2-benzyloxy carbonyl amino-3-methylbutanoic acid, d-valine, n-phenylmethoxy carbonyl |
PubChem CID | 712434 |
IUPAC Name | (2R)-3-methyl-2-(phenylmethoxycarbonylamino)butanoic acid |
SMILES | CC(C)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11332756
|
Thermo Scientific Alfa Aesar
L08856.03 |
1 g |
£21.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11342756
|
Thermo Scientific Alfa Aesar
L08856.06 |
5 g |
£60.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1685-33-2 | |
251.28 | |
CANZBRDGRHNSGZ-LLVKDONJSA-N | |
712434 | |
CC(C)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
C13H17NO4 | |
MFCD00065703 | |
z-d-val-oh, cbz-d-valine, n-benzyloxycarbonyl-d-valine, cbz-d-val-oh, n-cbz-d-valine, z-d-valine, n-carbobenzoxy-d-valine, benzyloxycarbonyl-d-valine, r-2-benzyloxy carbonyl amino-3-methylbutanoic acid, d-valine, n-phenylmethoxy carbonyl | |
(2R)-3-methyl-2-(phenylmethoxycarbonylamino)butanoic acid |
Specifications
1685-33-2 | |
C13H17NO4 | |
2056616 | |
CANZBRDGRHNSGZ-LLVKDONJSA-N | |
CC(C)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O | |
251.28 | |
251.28 | |
N-Benzyloxycarbonyl-D-valine |
58°C to 60°C | |
MFCD00065703 | |
z-d-val-oh, cbz-d-valine, n-benzyloxycarbonyl-d-valine, cbz-d-val-oh, n-cbz-d-valine, z-d-valine, n-carbobenzoxy-d-valine, benzyloxycarbonyl-d-valine, r-2-benzyloxy carbonyl amino-3-methylbutanoic acid, d-valine, n-phenylmethoxy carbonyl | |
+3.8° (c=2 in Acetic Acid) | |
(2R)-3-methyl-2-(phenylmethoxycarbonylamino)butanoic acid | |
712434 | |
≥98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only