missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-beta-alanine, 98%

Product Code. 15425568
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425568 5 g 5g
15435568 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425568 Supplier Thermo Scientific Alfa Aesar Supplier No. H59293.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2304-94-1 | C11H13NO4 | 223.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2304-94-1
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00037292
InChI Key GEVGRLPYQJTKKS-UHFFFAOYSA-N
Synonym z-beta-ala-oh, carbobenzyloxy-beta-alanine, n-cbz-beta-alanine, cbz-beta-alanine, n-benzyloxycarbonyl-beta-alanine, n-cbz-b-alanine, 3-benzyloxy carbonyl amino propanoic acid, cbz-beta-ala-oh, n-carbobenzoxy-beta-alanine
PubChem CID 75313
IUPAC Name 3-(phenylmethoxycarbonylamino)propanoic acid
SMILES OC(=O)CCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 105°C to 107°C
Linear Formula C6H5CH2OCONHCH2CH2CO2H
Beilstein 1882542
Quantity 5 g
Formula Weight 223.23
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-beta-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.