missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-4-hydroxy-L-proline, 99%

Product Code. 15476318
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476318 5 g 5g
15486318 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476318 Supplier Thermo Scientific Alfa Aesar Supplier No. H59882.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13504-85-3 | C13H15NO5 | 265.27 g/mol

A competitive inhibitor of porcine kidney prolidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A competitive inhibitor of porcine kidney prolidase.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13504-85-3
Molecular Formula C13H15NO5
Molecular Weight (g/mol) 265.27
MDL Number MFCD00037329,MFCD00037329,MFCD00238423,MFCD00037329,MFCD00238423,MFCD03428474
InChI Key WWVCWLBEARZMAH-UHFFFAOYNA-N
Synonym 2s-1-benzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, z-l-4-hydroxyproline, n-cbz-4-hydroxyproline, n-cbz-4-hydroxy-l-proline, n-benzyloxycarbonyl-4-hydroxyproline, n-carbobenzyloxy-4-hydroxy-l-proline, 4-hydroxy-n-benzyloxycarbonyl-l-proline, 1-carbobenzyloxy-4-hydroxy-l-proline, s-1-cbz-4-hydroxy-2-pyrrolidinecarboxylic acid
PubChem CID 12731799
SMILES OC1CC(N(C1)C(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 104°C to 107°C
Quantity 5 g
Beilstein 90295
Solubility Information Slightly soluble in water.
Optical Rotation −54° (c=2 in Ethanol)
IUPAC Name (2S)-4-hydroxy-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
Formula Weight 265.26
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-4-hydroxy-L-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.