missing translation for 'onlineSavingsMsg'
Learn More

N-[(Benzyloxy)carbonyl]glycylglycine, TRC

Product Code. 30346987
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346987 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346987 Supplier Toronto Research Chemicals Supplier No. TRCB287100250MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(Benzyloxy)carbonyl]glycylglycine
CAS 2566-19-0
Molecular Formula C12H14N2O5
Quantity 250 mg
Synonym (Benzyloxycarbonyl)glycylglycine, (Carbobenzoxy)glycylglycine, (Carbobenzyloxy)glycylglycine, N-(Benzyloxycarbonyl)glycylglycine, N- (Benzyloxycarbonylaminoacetyl)glycine, N-(Carbobenzoxy)glycylglycine, N-(Carbobenzyloxy)glycylglycine, N-(N-Cbz-glycyl)glycine, N-[N- (Benzyloxycarbonyl)glycyl]glycine, NSC 29727
InChI Formula InChI=1S/C12H14N2O5/c15-10(13-7-11(16)17)6-14-12(18)19-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,13,15)(H,14,18)(H,16,17)
SMILES O=C(O)CNC(CNC(OCC1=CC=CC=C1)=O)=O
IUPAC Name ((benzyloxy)carbonyl)glycylglycine
Molecular Weight (g/mol) 266.25
Formula Weight 266.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.