missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzylnaltrindole hydrochloride, Tocris Bioscience™
Selective non-peptide δ2 antagonist
Chemical Identifiers
CAS | 1206487-81-1 |
---|---|
Molecular Formula | C33H33ClN2O3 |
Molecular Weight (g/mol) | 541.088 |
InChI Key | KDEBSUYWJUBPCB-REBVNRAYSA-N |
Synonym | n-benzylnaltrindole hydrochloride |
PubChem CID | 131845649 |
SMILES | C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6CC9=CC=CC=C9.Cl |
Description
Potent δ2-selective opioid receptor antagonist with a long duration of action in vivo.Chemical Identifiers
1206487-81-1 | |
541.088 | |
n-benzylnaltrindole hydrochloride | |
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6CC9=CC=CC=C9.Cl |
C33H33ClN2O3 | |
KDEBSUYWJUBPCB-REBVNRAYSA-N | |
131845649 |
Specifications
1206487-81-1 | |
n-benzylnaltrindole hydrochloride | |
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6CC9=CC=CC=C9.Cl | |
131845649 | |
>98% |
C33H33ClN2O3 | |
KDEBSUYWJUBPCB-REBVNRAYSA-N | |
541.088 | |
541.09 | |
N-Benzylnaltrindole hydrochloride |
Safety and Handling
Recommended Storage : Store at Room Temperature