missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylidenemethylamine, 99%

Product Code. 10705234 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10705234 5 g 5g
10735171 25 g 25g
10656862 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10705234 Supplier Thermo Scientific Acros Supplier No. 106010050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-29-7 | C8H9N | 119.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-29-7
MDL Number MFCD00008294
InChI Key HXTGGPKOEKKUQO-UHFFFAOYSA-N
Synonym n-benzylidenemethylamine, n-benzylidenemethanamine, benzylidenemethylamine, n-methylbenzaldimine, methanamine, n-phenylmethylene, benzylidenemethanamine, n-methylbenzylideneamine, methylamine, n-benzylidene, n-phenylmethylene methanamine, benzylidene-methyl-amine
PubChem CID 73954
IUPAC Name N-methyl-1-phenylmethanimine
SMILES CN=CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Density 96%g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H9N
Linear Formula C6H5CH=NCH3
Refractive Index 1.5500 to 1.5540 (20°C, 589nm)
Quantity 5 g
Beilstein 07, 213
Molecular Weight (g/mol) 119.167
Formula Weight 119.17
Percent Purity ≥98.5% (GC)
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material N-Benzylidenemethylamine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Benzylidenemethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.