missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylideneaniline, 99%

Product Code. 15413097
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413097 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413097 Supplier Thermo Scientific Alfa Aesar Supplier No. A14746.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 538-51-2 | C13H11N | 181.24 g/mol

N-Benzylideneaniline is an additional stain which is used in various immunoassays including flow cytometry, immunofluorescense, immunohistochemistry, and other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylideneaniline is an additional stain which is used in various immunoassays including flow cytometry, immunofluorescense, immunohistochemistry, and other applications.

Solubility
Soluble in water (partly), methanol, chloroform, alcohol, and acetic anhydride.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 538-51-2
Melting Point 52°C to 54°C
Flash Point >110°C
Molecular Formula C13H11N
MDL Number MFCD00003027
Quantity 5 g
Merck Index 14,1138
Synonym n-benzylideneaniline, benzalaniline, benzylideneaniline, e-n-benzylideneaniline, n-benzylidenaniline
Solubility Information Soluble aq. soln. (partly),methanol,chloroform,alcohol,and acetic anhydride.
InChI Key UVEWQKMPXAHFST-UHFFFAOYSA-N
SMILES C(=NC1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name N,1-diphenylmethanimine
Molecular Weight (g/mol) 181.24
PubChem CID 10858
Formula Weight 181.24
Percent Purity 99%
Chemical Name or Material N-Benzylideneaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.