missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylideneaniline, 99%

Product Code. 11439120
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439120 25 g 25g
11449120 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439120 Supplier Thermo Scientific Alfa Aesar Supplier No. A14746.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 538-51-2 | C13H11N | 181.24 g/mol

N-Benzylideneaniline is an additional stain which is used in various immunoassays including flow cytometry, immunofluorescense, immunohistochemistry, and other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylideneaniline is an additional stain which is used in various immunoassays including flow cytometry, immunofluorescense, immunohistochemistry, and other applications.

Solubility
Soluble in water (partly), methanol, chloroform, alcohol, and acetic anhydride.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-51-2
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.24
MDL Number MFCD00003027
InChI Key UVEWQKMPXAHFST-UHFFFAOYSA-N
Synonym n-benzylideneaniline, benzalaniline, benzylideneaniline, e-n-benzylideneaniline, n-benzylidenaniline, n-benzalaniline, benzaldehyde anil, aniline, n-benzylidene, benzenamine, n-phenylmethylene, n-phenylmethylene benzenamine
PubChem CID 10858
IUPAC Name N,1-diphenylmethanimine
SMILES C(=NC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 52°C to 54°C
Flash Point >110°C (230°F)
Quantity 25 g
Merck Index 14,1138
Solubility Information Soluble in water (partly),methanol,chloroform,alcohol,and acetic anhydride.
Formula Weight 181.24
Percent Purity 99%
Chemical Name or Material N-Benzylideneaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.