missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylethanolamine, TRC

Product Code. 30329293
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329293 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329293 Supplier Toronto Research Chemicals Supplier No. TRCB276445250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzylethanolamine
CAS 104-63-2
Molecular Formula C9H13NO
Quantity 250 g
Synonym 2-(N-Benzylamino)ethanol, 2-[(Phenylmethyl)amino]ethanol, Benzyl(2-hydroxyethyl)amine, Benzylaminoethanol, Benzylethanolamine, N-(2-Hydroxyethyl)benzenemethanamine, N-(2-Hydroxyethyl)benzylamine, N-Benzyl-2-aminoethanol, N-Benzyl-2-hydroxyethylamine, N-Benzylaminoethanol, N-Benzylethanolamine, NSC 11271, NSC 177008, NSC 60267
InChI Formula InChI=1S/C9H13NO/c11-7-6-10-8-9-4-2-1-3-5-9/h1-5,10-11H,6-8H2
SMILES OCCNCc1ccccc1
IUPAC Name 2-(benzylamino)ethanol
Molecular Weight (g/mol) 151.21
Formula Weight 151.0997
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.