Learn More
N-Benzyldimethylamine, 98+%, Thermo Scientific Chemicals
Description
N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.
Solubility
Slightly miscible with water.
Notes
Air sensitive. Incompatible with strong acids, strong bases, strong oxidizing agents, strong reducing agents and carbon dioxide.
Specifications
Melting Point | -75°C |
Density | 0.9 |
Boiling Point | 183°C to 184°C |
Flash Point | 54°C (129°F) |
Linear Formula | C6H5CH2N(CH3)2 |
Refractive Index | 1.501 |
Quantity | 5000 mL |
UN Number | UN2619 |
Beilstein | 1099620 |
Sensitivity | Air sensitive |
Show More |
RUO – Research Use Only