missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl Glycine Hydrochloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | N-Benzyl Glycine Hydrochloride |
| CAS | 7689-50-1 |
| Molecular Formula | C9H11NO2 . HCl |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C9H11NO2.ClH/c11-9(12)7-10-6-8-4-2-1-3-5-8;/h1-5,10H,6-7H2,(H,11,12);1H |
| SMILES | Cl.OC(=O)CNCc1ccccc1 |
| IUPAC Name | 2-(benzylamino)acetic acid;hydrochloride |
| Molecular Weight (g/mol) | 201.65 |
| Formula Weight | 201.0557 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction