missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl Glycine Hydrochloride, TRC

Product Code. 30342699
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30342699

Brand: Toronto Research Chemicals TRCB27980850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyl Glycine Hydrochloride
CAS 7689-50-1
Molecular Formula C9H11NO2 . HCl
Quantity 50 mg
InChI Formula InChI=1S/C9H11NO2.ClH/c11-9(12)7-10-6-8-4-2-1-3-5-8;/h1-5,10H,6-7H2,(H,11,12);1H
SMILES Cl.OC(=O)CNCc1ccccc1
IUPAC Name 2-(benzylamino)acetic acid;hydrochloride
Molecular Weight (g/mol) 201.65
Formula Weight 201.0557
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.