missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl Epinephrine, TRC

Product Code. 17941215
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941215 10 mg 10mg
17933535 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17941215 Supplier Toronto Research Chemicals Supplier No. TRCB27640010MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1095714-91-2
Molecular Formula C16 H19 N O3
Molecular Weight (g/mol) 273.33
Synonym rac Adrenaline EP Impurity D
IUPAC Name 4-[2-[benzyl(methyl)amino]-1-hydroxy-ethyl]benzene-1,2-diol
SMILES CN(CC(O)c1ccc(O)c(O)c1)Cc2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H19NO3/c1-17(10-12-5-3-2-4-6-12)11-16(20)13-7-8-14(18)15(19)9-13/h2-9,16,18-20H,10-11H2,1H3
Formula Weight 273.1365
Chemical Name or Material N-Benzyl Epinephrine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.