missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-piperidone, 99%

Product Code. 10409400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Glass bottle
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10409400 250 mL Glass bottle 250mL
10723831 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10409400 Supplier Thermo Scientific Acros Supplier No. 106132500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3612-20-2 | C12H15NO | 189.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3612-20-2
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.26
MDL Number MFCD00006192
InChI Key SJZKULRDWHPHGG-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-one
PubChem CID 19220
IUPAC Name 1-benzylpiperidin-4-one
SMILES C1CN(CCC1=O)CC2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Straw
Density 1.0600g/mL
Boiling Point 133.0°C to 135.0°C (7.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5390 to 1.5410
Quantity 250 mL
Beilstein 21,IV,3189
Specific Gravity 1.06
Solubility Information Solubility in water: 12g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, d.m.f.,, acetone and chloroform
Formula Weight 189.26
Percent Purity 99%
Physical Form Oily Liquid
Chemical Name or Material N-Benzyl-4-piperidone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Benzyl-4-piperidone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.