missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-chloropentanamide, TRC

Product Code. 30331280
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 30331280

Brand: Toronto Research Chemicals TRCB23363550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyl-4-chloropentanamide
Analyte or Component Names N-Benzyl-4-chloropentanamide
Shipping Condition Room Temperature
Molecular Formula C3 H8 Br O3 P
InChI Formula InChI=1S/C3H8BrO3P/c4-2-1-3-8(5,6)7/h1-3H2,(H2,5,6,7)
Solution Type Neat
SMILES OP(=O)(O)CCCBr
IUPAC Name 3-bromopropylphosphonic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 202.972
Quantity 50 mg
Formula Weight 201.939
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.