missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl-3-(tert-butyldimethylsilanyloxymethyl)piperid-4-one, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 333.54 |
Percent Purity | 95% |
Chemical Name or Material | N-Benzyl-3-(tert-butyldimethylsilanyloxymethyl)piperid-4-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'