missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-2-pyridineethylamine, Thermo Scientific™

Product Code. 11449390
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11449390 1 g 1g
11439390 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 11449390

Brand: Thermo Scientific Alfa Aesar H50257.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6312-25-0
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00089263
InChI Key WUDUNBALAGWKQB-UHFFFAOYSA-N
Synonym n-benzyl-2-pyridin-2-yl ethanamine, n-benzyl-2-pyridin-2-yl-ethanamine, benzyl 2-pyridin-2-yl ethyl amine, n-benzyl-2-pyridin-2-ylethylamine, benzyl 2-2-pyridyl ethyl amine, acmc-20ak6u, n-benzyl-2-pyridineethylamine, 2-2-benzylaminoethyl pyridine, benzyl-2-pyridin-2-yl-ethyl amine, n-phenylmethyl-2-pyridin-2-ylethanamine
PubChem CID 238354
IUPAC Name N-benzyl-2-pyridin-2-ylethanamine
SMILES C1=CC=C(C=C1)CNCCC2=CC=CC=N2

Specifications

Quantity 1 g
UN Number UN2735
Formula Weight 212.3
Chemical Name or Material N-Benzyl-2-pyridineethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.