missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzoylcytidine, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | N-Benzoylcytidine |
| CAS | 13089-48-0 |
| Molecular Formula | C16 H17 N3 O6 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C16H17N3O6/c20-8-10-12(21)13(22)15(25-10)19-7-6-11(18-16(19)24)17-14(23)9-4-2-1-3-5-9/h1-7,10,12-13,15,20-22H,8H2,(H,17,18,23,24)/t10-,12-,13-,15-/m1/s1 |
| SMILES | OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=NC2=O)NC(=O)c3ccccc3 |
| IUPAC Name | N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide |
| Molecular Weight (g/mol) | 347.32 |
| Formula Weight | 347.1117 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction