missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoylaminopurine, 99%

Product Code. 11358085
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11358085

Brand: Thermo Scientific Alfa Aesar L08292.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4005-49-6 | C12H9N5O | 239.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4005-49-6
Molecular Formula C12H9N5O
Molecular Weight (g/mol) 239.238
MDL Number MFCD00037927
InChI Key QQJXZVKXNSFHRI-UHFFFAOYSA-N
Synonym n6-benzoyladenine, n-7h-purin-6-yl benzamide, n-benzoylaminopurine, n-9h-purin-6-yl benzamide, 6-benzoylaminopurine, benzamide, n-1h-purin-6-yl, 6-benzamidopurine, adenine,6n-benzoyl, phenyl-n-purin-6-ylcarboxamide, n-benzoyladenine
PubChem CID 97075
IUPAC Name N-(7H-purin-6-yl)benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=NC=NC3=C2NC=N3

Specifications

Melting Point 241°C to 244°C
Quantity 5 g
Beilstein 20585
Formula Weight 239.23
Percent Purity 99%
Chemical Name or Material N-Benzoylaminopurine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.