missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyladenosine, Thermo Scientific Chemicals

Product Code. 16594434 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16594434 5 g 5g
16584434 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16594434 Supplier Thermo Scientific Acros Supplier No. 464690050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4546-55-8
Molecular Formula C17H17N5O5
Molecular Weight (g/mol) 371.35
MDL Number MFCD00056988
InChI Key NZDWTKFDAUOODA-UCQOUBKUNA-N
IUPAC Name N-{9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12

Specifications

Melting Point 141.0°C to 145.0°C
Quantity 5 g
Formula Weight 371.35
Chemical Name or Material N-Benzoyladenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.