missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl Isothiocyanate, TRC

Product Code. 30343897
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. Quantity unitSize
30343897 10 mL 10mL
1 options
This item is not returnable. View return policy

Product Code. 30343897

Brand: Toronto Research Chemicals TRCB20819510ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzoyl Isothiocyanate
CAS 532-55-8
Molecular Formula C8H5NOS
Quantity 10 mL
Synonym Benzoylthiocarbimide, NSC 29259
InChI Formula InChI=1S/C8H5NOS/c10-8(9-6-11)7-4-2-1-3-5-7/h1-5H
SMILES O=C(N=C=S)c1ccccc1
IUPAC Name benzoyl isothiocyanate
Molecular Weight (g/mol) 163.2
Formula Weight 163.0092
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.