missing translation for 'onlineSavingsMsg'
Learn More

n-Amyl Alcohol, 99.5%, MP Biomedicals

Product Code. 11472845 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472845 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11472845 Supplier MP Biomedicals Supplier No. 0521105780
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 71-41-0
Molecular Formula C5H12O
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
Quantity 100 g
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
SMILES CCCCCO
IUPAC Name pentan-1-ol
Molecular Weight (g/mol) 88.15
PubChem CID 6276
ChEBI CHEBI:44884
Formula Weight 88.15
Chemical Name or Material n-Amyl Alcohol, 99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.