missing translation for 'onlineSavingsMsg'
Learn More

n-Amyl acetate, 99%

Product Code. 10114293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10114293 2.5 L Glass bottle 2.5 L
10011510 250 mL Glass bottle 250mL
10513891 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10114293 Supplier Thermo Scientific Acros Supplier No. 149180025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

n-Amyl acetate, 99%, C7H14O2, CAS Number-628-63-7, 1-pentyl acetate, acetic acid, pentyl ester, amyl acetate, amyl acetic ester, amyl acetic ether, birnenoel, n-amyl acetate, n-pentyl acetate, n-pentyl ethanoate, pent-acetate, 2.5L, 98.5% min. (GC), 02, 131, 149.0 deg.C, 100.0, CHEBI:87362 | CAS: 628-63-7 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009500
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
Synonym amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
PubChem CID 12348
ChEBI CHEBI:87362
IUPAC Name pentyl acetate
SMILES CCCCCOC(=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.8°C
Color Colorless
Density 0.8700g/mL
Boiling Point 149.0°C
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2(CH2)4CH3
Refractive Index 1.4010 to 1.4030
Quantity 2.5 L
Beilstein 02, 131
Specific Gravity 0.87
Solubility Information Solubility in water: 10g/L (20c). Other solubilities: miscible with alcohol and ether
Formula Weight 130.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Amyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Amyl acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.