missing translation for 'onlineSavingsMsg'
Learn More

N-Amyl Acetate, 99+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10142040 Shop All Fisher Chemical Products
Change view
Click to view available options
500mL, Amber glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10142040 500mL, Amber glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10142040 Supplier Fisher Chemical Supplier No. A/6800/08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 628-63-7
Melting Point -70.8°C
Color Colorless
Boiling Point 149°C
Packaging Amber glass bottle
Molecular Formula C7H14O2
MDL Number 9500
Quantity 500 mL
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
SMILES CCCCCOC(C)=O
IUPAC Name pentyl acetate
Molecular Weight (g/mol) 130.19
Formula Weight 130.19g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Amyl acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.