missing translation for 'onlineSavingsMsg'
Learn More

N-Amino-D-proline, TRC

Product Code. 30321097
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321097 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321097 Supplier Toronto Research Chemicals Supplier No. TRCA62800050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Amino-D-proline
CAS 10139-05-6
Molecular Formula C5 H10 N2 O2
Quantity 50 mg
InChI Formula InChI=1S/C5H10N2O2/c6-7-3-1-2-4(7)5(8)9/h4H,1-3,6H2,(H,8,9)/t4-/m1/s1
SMILES NN1CCC[C@@H]1C(=O)O
IUPAC Name (2R)-1-aminopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 130.15
Formula Weight 130.0742
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.