missing translation for 'onlineSavingsMsg'
Learn More

N-Amino-D-proline, TRC

Product Code. 30313572
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313572 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313572 Supplier Toronto Research Chemicals Supplier No. TRCA628000500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material N-Amino-D-proline
CAS 10139-05-6
Molecular Formula C5 H10 N2 O2
Quantity 500 mg
InChI Formula InChI=1S/C5H10N2O2/c6-7-3-1-2-4(7)5(8)9/h4H,1-3,6H2,(H,8,9)/t4-/m1/s1
SMILES NN1CCC[C@@H]1C(=O)O
IUPAC Name (2R)-1-aminopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 130.15
Formula Weight 130.0742
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.