missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N(alpha)-Fmoc-L-tryptophan, 98%

Product Code. 11474617
Change view
Click to view available options
Quantity:
5g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474617 5g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474617 Supplier Alfa Aesar™ Supplier No. B21130.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 35737-15-6
Molecular Formula C26H22N2O4
MDL Number MFCD00037126
Beilstein 4216624
Synonym fmoc-trp-oh, nalpha-fmoc-l-tryptophan, fmoc-l-tryptophan, n-fmoc-l-tryptophan, fmoc-l-trp-oh, fmoc-tryptophan, l-tryptophan, n-9h-fluoren-9-ylmethoxy carbonyl, n-9h-fluoren-9-ylmethoxy carbonyl-l-tryptophan, 9-fluorenylmethoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-indol-3-yl propanoic acid
InChI Key MGHMWKZOLAAOTD-DEOSSOPVSA-N
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CNC5=CC=CC=C54)C(=O)O
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 426.472
Quantity 5g
PubChem CID 978343
Formula Weight 426.48
Optical Rotation −27° (c=1 in DMF)
Percent Purity 98%
Chemical Name or Material N(alpha)-Fmoc-L-tryptophan
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.