missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-BOC-N(in)-Formyl-L-tryptophan, 97%, Thermo Scientific™

Product Code. 11985511 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11985511 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11985511 Supplier Thermo Scientific Acros Supplier No. 440910050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 47355-10-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 123.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Molecular Formula C17H20N2O5
MDL Number MFCD00065992
Quantity 5 g
Specific Rotation 19.7
Synonym boc-trp for-oh, n-tert-butoxycarbonyl-n'-formyl-l-tryptophan, chembl63415, nalpha-tert-butoxycarbonyl-n1-formyl-l-tryptophan, s-2-tert-butoxycarbonyl amino-3-1-formyl-1h-indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-formylindol-3-yl propanoic acid, boc-trp cho-oh, pubchem12191, t-boc-trp cho-oh, nalpha-boc-n1-formyl-l-tryptophan
InChI Key IHXHBYFWSOYYTR-ZDUSSCGKSA-N
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CN(C=O)C2=CC=CC=C12)C(O)=O
IUPAC Name (2S)-3-(1-formylindol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molecular Weight (g/mol) 332.36
PubChem CID 7017963
Formula Weight 332.36
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-α-BOC-N(in)-Formyl-L-tryptophan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.