missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Boc-N(epsilon),N(epsilon)-dimethyl-L-lysine, 97%

Product Code. 15449177
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449177 250 mg 250mg
15439177 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449177 Supplier Thermo Scientific Chemicals Supplier No. H52437.MD
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65671-53-6 | C13H26N2O4 | 274.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65671-53-6
Molecular Formula C13H26N2O4
Molecular Weight (g/mol) 274.36
MDL Number MFCD00076965
InChI Key KPXRFYHPDPDJSY-UEQNJFAPNA-N
Synonym boc-lys me 2-oh, l-lysine,n2-1,1-dimethylethoxy carbonyl-n6,n6-dimethyl, 2s-2-tert-butoxycarbonyl amino-6-dimethylamino hexanoic acid, boc-lys me-oh, nalpha-boc-nepsilon-dimethyl-l-lysine, n-alpha-boc-s-2-amino-6-dimethylamino hexanoic acid, 2 s-+-tert-butoxycarbonylamino-6-dimethylaminohexanoic acid, s-2-tert-butoxycarbonylamino-6-dimethylamino hexanoic acid, 2s-2-tert-butoxy carbonyl amino-6-dimethylamino hexanoic acid
PubChem CID 7018778
IUPAC Name (2S)-6-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CN(C)CCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 274.36
Percent Purity 97%
Chemical Name or Material N(alpha)-Boc-N(epsilon),N(epsilon)-dimethyl-L-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.