missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Boc-D-2,3-diaminopropionic acid, 97%

Product Code. 15469587
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469587 250 mg 250mg
15449587 1 g 1g
15459587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15469587 Supplier Thermo Scientific Alfa Aesar Supplier No. H52821.MD
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76387-70-7 | C8H16N2O4 | 204.226 g/mol

N(alpha)-Boc-D-2,3-diaminopropionic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N(alpha)-Boc-D-2,3-diaminopropionic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76387-70-7
Molecular Formula C8H16N2O4
Molecular Weight (g/mol) 204.226
MDL Number MFCD01632072
InChI Key KRJLRVZLNABMAT-RXMQYKEDSA-N
Synonym boc-d-dap-oh, boc-d-2,3-diaminopropionic acid, r-3-amino-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-amino-2-tert-butoxy carbonyl amino propanoic acid, d-alanine, 3-amino-n-1,1-dimethylethoxy carbonyl, 2r-3-amino-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-amino-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, ambotzbaa1179, pubchem14749, 3-amino-n-boc-d-alanine
PubChem CID 7020984
IUPAC Name (2R)-3-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CN)C(=O)O

Specifications

Melting Point 200°C to 203°C
Quantity 250 mg
Optical Rotation +14° (c=1 in Water)
Formula Weight 204.23
Percent Purity 97%
Chemical Name or Material N(alpha)-Boc-D-2,3-diaminopropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.