missing translation for 'onlineSavingsMsg'
Learn More

N-Allylmethylamine, 96%

Product Code. 11431850
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11431850

Brand: Thermo Scientific Alfa Aesar H27551.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-37-2 | C4H9N | 71.123 g/mol

N-Allylmethylamine is used in the production of N-allyl-alfa,alfa-dichloro-N-methylacetamide. It is used as a functional monomer to coordinate with Cd(II)/Zn(II) ions to get the poly methyl methacrylate -butyl methacrylate - N-allylmethylamine (PMBA) films, which is useful as building blocks to fabricate a photo-catalytic cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Allylmethylamine is used in the production of N-allyl-alfa,alfa-dichloro-N-methylacetamide. It is used as a functional monomer to coordinate with Cd(II)/Zn(II) ions to get the poly methyl methacrylate -butyl methacrylate - N-allylmethylamine (PMBA) films, which is useful as building blocks to fabricate a photo-catalytic cell.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-37-2
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.123
MDL Number MFCD00008641
InChI Key IOXXVNYDGIXMIP-UHFFFAOYSA-N
Synonym n-allylmethylamine, n-methylallylamine, 2-propen-1-amine, n-methyl, allylmethylamine, methylallylamine, n-methyl-2-propen-1-amine, n-allyl-n-methylamine, methyl prop-2-en-1-yl amine, allylamine, n-methyl-7ci,8ci, 2-propen-1-amine, n-methyl-9ci
PubChem CID 69391
IUPAC Name N-methylprop-2-en-1-amine
SMILES CNCC=C

Specifications

Density 0.748
Boiling Point 61°C to 62°C
Flash Point −8°C (−18°F)
Refractive Index 1.412
Quantity 5 g
UN Number UN2733
Solubility Information Miscible with water.
Formula Weight 71.12
Percent Purity 96%
Chemical Name or Material N-Allylmethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.