missing translation for 'onlineSavingsMsg'
Learn More

N-Allylaniline, 95%

Product Code. 10448050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10448050 100 g 100g
10541564 5 g 5g
10306120 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10448050 Supplier Thermo Scientific Acros Supplier No. 102871000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 589-09-3 | C9H11N | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-09-3
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008638
InChI Key LQFLWKPCQITJIH-UHFFFAOYSA-N
Synonym n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine
PubChem CID 68525
IUPAC Name N-prop-2-enylaniline
SMILES C=CCNC1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.9820g/mL
Boiling Point 218°C to 220°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula C6H5NHCH2CH=CH2
Refractive Index 1.5620 to 1.564
Quantity 100 g
Beilstein 12, 170
Specific Gravity 0.982
Formula Weight 133.19
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Allylaniline, 95%
Show More Show Less
compliance-icons
Product Identifier
  • N-Allylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.