missing translation for 'onlineSavingsMsg'
Learn More

N-acylmannosamine 1-dehydrogenase Antibody, HRP, Novus Biologicals™

Product Code. 18342581 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342581 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342581 Supplier Novus Biologicals Supplier No. NBP1775650.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

N-acylmannosamine 1-dehydrogenase Polyclonal specifically detects N-acylmannosamine 1-dehydrogenase in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cholesterol Oxidase
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate HRP
Dilution Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate
Gene Accession No. P12676
Gene Alias choA, choD, choM, choS, COD
Host Species Goat
Immunogen Cholesterol Oxidase (Microorganism).
Purification Method Delipidation and Defibrination
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Cholesterol Oxidase
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.