missing translation for 'onlineSavingsMsg'
Learn More

N-Acryloxysuccinimide, 99%

Product Code. 10527172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10527172 5 g Glass bottle 5g
10085702 1 g Glass bottle 1g
11976911 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10527172 Supplier Thermo Scientific Acros Supplier No. 400300050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 38862-24-7 | C7H7NO4 | 169.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38862-24-7
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00078261
InChI Key YXMISKNUHHOXFT-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl acrylate, n-acryloxysuccinimide, n-succinimidyl acrylate, acrylic acid n-hydroxysuccinimide ester, 1-acryloyloxy-2,5-pyrrolidinedione, n-acryloyloxysuccinimide, 2,5-pyrrolidinedione, 1-1-oxo-2-propenyl oxy, 2,5-dioxopyrrolidin-1-yl prop-2-enoate, poly n-acryloxysuccinimide
PubChem CID 181508
IUPAC Name (2,5-dioxopyrrolidin-1-yl) prop-2-enoate
SMILES C=CC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 69.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diethyl ether,hexane,ethyl acetate
Formula Weight 169.14
Percent Purity 99%
Physical Form Crystals and/or Chunks
Chemical Name or Material N-Acryloxysuccinimide
Show More Show Less
compliance-icons
Product Identifier
  • N-Acryloxysuccinimide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.