missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyltyrosine (N-Acetyl-L-Tyrosine), Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
N-Acetyltyrosine (N-Acetyl-L-Tyrosine)
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-L-tyrosine |
| CAS | 537-55-3 |
| Analyte or Component Names | N-Acetyl-L-tyrosine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H13 N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-Acetyltyrosine, (2S)-2-(Acetylamino)-3-(4-hydroxyphenyl)propanoic acid, Tyrosine, N-acetyl-, L- (6CI,8CI), (+)-(2S)-2-(Acetylamino)-3-(4-hydroxyphenyl)propanoic acid, (2S)-2-Acetylamino-3-(4-hydroxyphenyl)propanoic acid, L-N-Acetyltyrosine, N-Acetyl-L-tyrosine, N-Acetyltyrosine, NSC 10853, Tyr-Excel |
| InChI Formula | InChI=1S/C11H13NO4/c1-7(13)12-10(11(15)16)6-8-2-4-9(14)5-3-8/h2-5,10,14H,6H2,1H3,(H,12,13)(H,15,16)/t10-/m0/s1 |
| SMILES | CC(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)O |
| IUPAC Name | (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction