missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylputrescine hydrochloride, MedChemExpress
Description
N-Acetylputrescine hydrochloride is a putrescine derivative.
Specifications
Specifications
| Chemical Name or Material | N-Acetylputrescine hydrochloride |
| Physical Form | Powder |
| CAS | 18233-70-0 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C6H15ClN2O |
| Solubility Information | DMSO : 250 mg/mL (1500.15 mM; Need ultrasonic) ∣H2O : 200 mg/mL (1200.12 mM; Need ultrasonic) |
| SMILES | CC(NCCCCN)=O.Cl |
| Molecular Weight (g/mol) | 166.65 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?