missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylmorpholine, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | N-Acetylmorpholine |
| CAS | 1696-20-4 |
| Molecular Formula | C6H11NO2 |
| Quantity | 500 mg |
| Synonym | 1-(4-Morpholinyl)ethanone (ACI), Morpholine, 4-acetyl- (6CI, 7CI, 8CI, 9CI), 1-(Morpholin-4-yl)ethan-1-one, 1-(Morpholin-4-yl)ethanone, 1-Morpholinoethan-1-one, 1-Morpholinoethanone, 4-Acetylmorpholine, Acetomorpholide, N-Acetylmorpholine, NSC 2764 |
| InChI Formula | InChI=1S/C6H11NO2/c1-6(8)7-2-4-9-5-3-7/h2-5H2,1H3 |
| SMILES | CC(=O)N1CCOCC1 |
| IUPAC Name | 1-morpholin-4-ylethanone |
| Molecular Weight (g/mol) | 129.16 |
| Formula Weight | 129.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction