missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylmorpholine, 98%

Product Code. 11471127
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11471127

Brand: Thermo Scientific Alfa Aesar B24120.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1696-20-4 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1696-20-4
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00006171
InChI Key KYWXRBNOYGGPIZ-UHFFFAOYSA-N
Synonym 4-acetylmorpholine, n-acetylmorpholine, 1-morpholinoethanone, morpholine, 4-acetyl, n-acetylmorfolin, n-acetylmorfolin czech, ethanone, 1-4-morpholinyl, n-acetyl morpholine, 1-morpholin-4-yl ethan-1-one, acetylmorpholine
PubChem CID 15543
IUPAC Name 1-morpholin-4-ylethanone
SMILES CC(=O)N1CCOCC1

Specifications

Melting Point 9°C to 10°C
Density 1.116
Boiling Point 245°C to 246°C
Flash Point >110°C (230°F)
Refractive Index 1.4845
Quantity 25 g
Beilstein 111971
Formula Weight 129.16
Percent Purity 98%
Chemical Name or Material N-Acetylmorpholine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.