missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylmannosamine Kinase/GNE Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33260-100ul
This item is not returnable.
View return policy
Description
N-Acetylmannosamine Kinase/GNE Monoclonal antibody specifically detects N-Acetylmannosamine Kinase/GNE in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| N-Acetylmannosamine Kinase/GNE | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, DMRV, GLCNENM, glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase, IBM2, N-acylmannosamine kinase, Uae1, UDP-GlcNAc-2-epimerase/ManAc kinase, UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase | |
| A synthetic peptide corresponding to a sequence within amino acids 623-722 of human N-Acetylmannosamine Kinase/GNE (Q9Y223).,, Sequence:, GALHLIQAAKLGNAKAQSILRTAGTALGLGVVNILHTMNPSLVILSGVLASHYIHIVKDVIRQQALSSVQDVDVVVSDLVDPALLGAASMVLDYTTRRIY | |
| 100 μL | |
| Cardiovascular Biology, Cell Biology, Signal Transduction | |
| 10020 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction