missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylglucosaminyltransferase III/MGAT3 Antibody, Novus Biologicals™

Product Code. 18326157 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326157 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326157 Supplier Novus Biologicals Supplier No. H00004248D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

N-Acetylglucosaminyltransferase III/MGAT3 Polyclonal antibody specifically detects N-Acetylglucosaminyltransferase III/MGAT3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen N-Acetylglucosaminyltransferase III/MGAT3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. AAH75025.1
Gene Alias beta-1,4-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase, EC 2.4.1.144, FLJ43371, GGNT3, GlcNAc-T III, GNT3, GNT-IIIMGC142278, mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, MGC141943, N-acetylglucosaminyltransferase III, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase III
Host Species Rabbit
Immunogen MGAT3 (AAH75025.1, 1 a.a. - 533 a.a.) full-length human protein. MKMRRYKLFLMFCMAGLCLISFLHFFKTLSYVTFPRELASLSPNLVSSFFWNNAPVTPQASPEPGGPDLLRTPLYSHSPLLQPLPPSKAAEELHRVDLVLPEDTTEYFVRTKAGGVCFKPGTKMLERPPPGRPEEKPEGANGSSARRPPRYLLSARERTGGRGARRKWVECVCLPGWHGPSCGVPTVVQYSNLPTKERLVPREVPRRVINAINVNHEFDLLDVRFHELGDVVDAFVVCESNFTAYGEPRPLKFREMLTNGTFEYIRHKVLYVFLDHFPPGGRQDGWIADDYLRTFLTQDGVSRLRNLRPDDVFIIDDADEIPARDGVLFLKLYDGWTEPFAFHMRKSLYGFFWKQPGTLEVVSGCTVDMLQAVYGLDGIRLRRRQYYTMPNFRQYENRTGHILVQWSLGSPLHFAGWHCSWCFTPEGIYFKLVSAQNGDFPRWGDYEDKRDLNYIRGLIRTGGWFDGTQQEYPPADPSEHMYAPKYLLKNYDRFHYLLDNPYQEPRSTAAGGWRHRGPEGRPPARGKLDEAEV
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4248
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.