missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ N-Acetylcysteine amide
GREENER_CHOICE

Product Code. p-7110849
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18747668

Brand: Tocris Bioscience™ 5619/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glutathione (GSH) precursor; neuroprotective

N-Acetylcysteine amide is a glutathione (GSH) precursor and cell-permeable antioxidant. N-Acetylcysteine amide replenishes intracellular Glutathione (GSH). N-Acetylcysteine amide directly reduces intracellular Glutathione disulfide (GSSG) to GSH without glutathione peroxidase. N-Acetylcysteine amide has anti-inflammatory activity through regulation of activation of NF-κB and HIF-1α, as well as modulation of reactive oxygen species. N-Acetylcysteine amide improves neuronal mitochondrial bioenergetics, reduces tissue damage and enhances functional recovery following spinal cord injury in rats. N-Acetylcysteine amide also enhances behavioral recovery in rats following traumatic brain injury. Neuroprotective.

For more information about how N-Acetylcysteine amide may be used, see our protocol: 3D Culture of Lung Alveolar Cells

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-(Acetylamino)-3-mercaptopropanamide
CAS 38520-57-9
Quantity 10 mg
Target Additional Antioxidant Compounds
Molecular Formula C5H10N2O2S
Formula Weight 162.21
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.